2-(1-Piperidinylmethyl)azepane 95%

Lieferant: Apollo Scientific

OR906041-1G
APOSOR906041-1GEA 348 EUR
APOSOR906041-1G
2-(1-Piperidinylmethyl)azepane 95%
2-(1-Piperidinylmethyl)azepane
Formel: C₁₂H₂₄N₂
Molecular Weight: 196,332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663744
CAS-Nummer: 383129-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR