7-Ethoxy-2-naphthalenol 95%

Lieferant: Apollo Scientific

OR906202-1G
APOSOR906202-1GEA 0 EUR
APOSOR906202-1G
7-Ethoxy-2-naphthalenol 95%
7-Ethoxy-2-naphthalenol
Formel: C₁₂H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11110689
CAS-Nummer: 57944-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR