2-Amino-4-methoxybenzonitril 98%

Lieferant: Apollo Scientific

OR906314-5G
APOSOR906314-5GEA 72.5 EUR
APOSOR906314-5G
2-Amino-4-methoxybenzonitril 98%
2-Amino-4-methoxybenzonitril
Formel: C₈H₈N₂O
Molecular Weight: 148.1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05149246
CAS-Nummer: 38487-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR