2-Amino-N-methyl-DL-propanamide 98%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-N-methylpropanamide
OR906370-1G
OR906370-5G
APOSOR906370-1GEA
114
EUR
APOSOR906370-1G
APOSOR906370-5G
2-Amino-N-methyl-DL-propanamide 98%
2-Amino-N-methyl-DL-propanamide
Formel:
C₄H₁₀N₂O Molecular Weight: 102.135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09927582 CAS-Nummer: 32012-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...