[4-(Dimethoxymethyl)pyridin-2-yl]boronic acid 95%
Lieferant: Apollo Scientific
OR906381-1G
APOSOR906381-1GEA
864
EUR
APOSOR906381-1G
[4-(Dimethoxymethyl)pyridin-2-yl]boronic acid 95%
[4-(Dimethoxymethyl)pyridin-2-yl]boronic acid
Formel:
C₈H₁₂BNO₄ Molecular Weight: 196,996 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23699491 CAS-Nummer: 1908439-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...