4-Bromo-7H-pyrrolo[2,3-d]pyrimidine 98%

Lieferant: Apollo Scientific

OR906413-5G
APOSOR906413-5GEA 151 EUR
APOSOR906413-5G
4-Bromo-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Bromo-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₆H₄BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08445698
CAS-Nummer: 889939-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR