tert-Butyl N-(furan-2-ylmethyl)carbamate 96%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (furan-2-ylmethyl)carbamate

OR906460-1G
APOSOR906460-1GEA 241 EUR
APOSOR906460-1G
tert-Butyl N-(furan-2-ylmethyl)carbamate 96%
tert-Butyl N-(furan-2-ylmethyl)carbamate
Formel: C₁₀H₁₅NO₃
Molecular Weight: 197,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00734859
CAS-Nummer: 178918-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR