4-(4-Chlorophenyl)-1,3-thiazole 98%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Chlorophenyl)thiazole

OR906578-1G
APOSOR906578-1GEA 60.9 EUR
APOSOR906578-1G
4-(4-Chlorophenyl)-1,3-thiazole 98%
4-(4-Chlorophenyl)-1,3-thiazole
Formel: C₉H₆ClNS
Molecular Weight: 195,669 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02665933
CAS-Nummer: 1826-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR