Ethyl 2-isocyanopropanoate 95%

Lieferant: Apollo Scientific

OR906638-100MG
APOSOR906638-100MGEA 952 EUR
APOSOR906638-100MG
Ethyl 2-isocyanopropanoate 95%
Ethyl 2-isocyanopropanoate
Formel: C₆H₉NO₂
Molecular Weight: 127.141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664649
CAS-Nummer: 33140-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR