3-Formyl-2-methylphenylboronic acid 96%

Lieferant: Apollo Scientific
Achtung

OR906707-1G
APOSOR906707-1GEA 679 EUR
APOSOR906707-1G
3-Formyl-2-methylphenylboronic acid 96%
3-Formyl-2-methylphenylboronic acid
Formel: C₈H₉BO₃
Molecular Weight: 163,966 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23380480
CAS-Nummer: 631909-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR