4-Bromchinazolin 97%

Lieferant: Apollo Scientific
Achtung

OR906785-250MG
APOSOR906785-250MGEA 79.9 EUR
APOSOR906785-250MG
4-Bromchinazolin 97%
4-Bromoquinazoline
Formel: C₈H₅BrN₂
Molecular Weight: 209,04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10697879
CAS-Nummer: 354574-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR