Pyrrolidin-3-ol hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(±)-3-Hydroxypyrrolidine hydrochloride
OR9068-25G
APOSOR9068-25GEA
176
EUR
APOSOR9068-25G
Pyrrolidin-3-ol hydrochloride 97%
Pyrrolidin-3-ol hydrochloride
Formel:
C4H10ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001754 CAS-Nummer: 86070-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...