2-Methyl-1-(naphthalen-2-yl)propan-2-amine 95%

Lieferant: Apollo Scientific

OR906807-250MG
APOSOR906807-250MGEA 574 EUR
APOSOR906807-250MG
2-Methyl-1-(naphthalen-2-yl)propan-2-amine 95%
2-Methyl-1-(naphthalen-2-yl)propan-2-amine
Formel: C₁₄H₁₇N
Molecular Weight: 199,3 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11519113
CAS-Nummer: 198226-63-0
UN: 2920
ADR: 8+3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR