2-Chlor-5-methoxy-4-pyridinboronsäure 98%
Lieferant: Apollo Scientific
OR906834-5G
APOSOR906834-5GEA
252
EUR
APOSOR906834-5G
2-Chlor-5-methoxy-4-pyridinboronsäure 98%
2-Chloro-5-methoxy-4-pyridineboronic acid
Formel:
C₆H₇BClNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09972106 CAS-Nummer: 1072945-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...