2-(Methoxymethyl)piperidine 95%

Lieferant: Apollo Scientific

OR906842-100MG OR906842-1G OR906842-250MG
APOSOR906842-100MGEA 190 EUR
APOSOR906842-100MG APOSOR906842-1G APOSOR906842-250MG
2-(Methoxymethyl)piperidine 95%
2-(Methoxymethyl)piperidine
Formel: C₇H₁₅NO
Molecular Weight: 129.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663567
CAS-Nummer: 104678-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR