1-Methylpyrrol-3-carbonsäure 97%
Lieferant: Apollo Scientific
Synonyme:
1-Methylpyrrole-3-carboxylic Acid
OR906844-5G
APOSOR906844-5GEA
253
EUR
APOSOR906844-5G
1-Methylpyrrol-3-carbonsäure 97%
1-Methylpyrrole-3-carboxylic acid
Formel:
C₆H₇NO₂ Molecular Weight: 125.1300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111733 CAS-Nummer: 36929-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...