2-Amino-5-bromo-4-methylpyrimidine 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Amino-4-methyl-5-bromopyrimidine

OR906853-1G
APOSOR906853-1GEA 21.9 EUR
APOSOR906853-1G
2-Amino-5-bromo-4-methylpyrimidine 98%
2-Amino-5-bromo-4-methylpyrimidine
Formel: C₅H₆BrN₃
Molecular Weight: 188.03 g/mol
Schmelzpunkt: 192…194 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00094469
CAS-Nummer: 17321-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR