Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid methyll ester, Methyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate

OR906870-1G OR906870-25G OR906870-100G OR906870-BULK
APOSOR906870-1GEA 14.7 EUR
APOSOR906870-1G APOSOR906870-25G APOSOR906870-100G APOSOR906870-BULK
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate 97%
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate

Bitte beachten Use only outdoors or in a well-ventilated area. Avoid breathing dust/fumes. Avoid release to the environment. Wear protective gloves. Wash all exposed external body areas thoroughly after handling. If in eyes rinse cautiously with hazard eye protection and face protection.

Formel: C₁₈H₂₈O₃
Schmelzpunkt: 60…67 °C
MDL: MFCD00210461
CAS-Nummer: 6386-38-5
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Purity 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR