6-Propoxypyridazin-3-amine 95%

Lieferant: Apollo Scientific

OR906932-1G
APOSOR906932-1GEA 91.5 EUR
APOSOR906932-1G
6-Propoxypyridazin-3-amine 95%
6-Propoxypyridazin-3-amine
Formel: C₇H₁₁N₃O
Molecular Weight: 153.1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835392
CAS-Nummer: 90008-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR