2-Ethoxy-4-pyridinboronsäure 97%

Lieferant: Apollo Scientific

OR906934-1G
APOSOR906934-1GEA 364 EUR
APOSOR906934-1G
2-Ethoxy-4-pyridinboronsäure 97%
2-Ethoxy-4-pyridineboronic acid
Formel: C₇H₁₀BNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07374989
CAS-Nummer: 1072946-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR