Methyl 3-tert-butylisoxazole-5-carboxylate 95%

Lieferant: Apollo Scientific

OR906938-100MG OR906938-1G OR906938-250MG
APOSOR906938-100MGEA 190 EUR
APOSOR906938-100MG APOSOR906938-1G APOSOR906938-250MG
Methyl 3-tert-butylisoxazole-5-carboxylate 95%
Methyl 3-tert-butylisoxazole-5-carboxylate
Formel: C₉H₁₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08752835
CAS-Nummer: 133674-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR