cis-4-Cyclohexen-1,2-dicarbonsäure 95%

Lieferant: Apollo Scientific

OR906968-500G
APOSOR906968-500GEA 102 EUR
APOSOR906968-500G
cis-4-Cyclohexen-1,2-dicarbonsäure 95%
cis-4-Cyclohexen-1,2-dicarbonsäure
Formel: C₈H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00070482
CAS-Nummer: 2305-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR