7-Chloro-1H-indole-2-carboxylic acid 97%

Lieferant: Apollo Scientific

OR906974-5G
APOSOR906974-5GEA 132 EUR
APOSOR906974-5G
7-Chloro-1H-indole-2-carboxylic acid 97%
7-Chloro-1H-indole-2-carboxylic acid
Formel: C₉H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02664428
CAS-Nummer: 28899-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR