1-(Dimethylamino)-2-methylpentan-3-one 95%

Lieferant: Apollo Scientific

OR906981-25G
APOSOR906981-25GEA 63.8 EUR
APOSOR906981-25G
1-(Dimethylamino)-2-methylpentan-3-one 95%
1-(Dimethylamino)-2-methylpentan-3-one
Formel: C₈H₁₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13185946
CAS-Nummer: 51690-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR