(5,6-Dimethoxypyridin-3-yl)boronic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dimethoxypyridine-5-boronic acid

OR906990-1G
APOSOR906990-1GEA 623 EUR
APOSOR906990-1G
(5,6-Dimethoxypyridin-3-yl)boronic acid 95%
(5,6-Dimethoxypyridin-3-yl)boronic acid
Formel: C₇H₁₀BNO₄
Molecular Weight: 182,97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18254414
CAS-Nummer: 1346526-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR