4-Chloro-5-methylpyrimidine 98%
Lieferant: Apollo Scientific
OR907028-5G
APOSOR907028-5GEA
212
EUR
APOSOR907028-5G
4-Chloro-5-methylpyrimidine 98%
4-Chloro-5-methylpyrimidine
Formel:
C₅H₅ClN₂ Molecular Weight: 128.5596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08236809 CAS-Nummer: 51957-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...