(1S,E)-(-)-Camphorquinone 3-oxime 95%

Lieferant: Apollo Scientific

OR907065-1G
APOSOR907065-1GEA 0 EUR
APOSOR907065-1G
(1S,E)-(-)-Camphorquinone 3-oxime 95%
(1S,E)-(-)-Camphorquinone 3-oxime
Formel: C₁₀H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00142700
CAS-Nummer: 251645-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR