(2,6-Dibromopyridin-4-yl)boronic acid > 98%
Lieferant: Apollo Scientific
OR907072-1G
APOSOR907072-1GEA
78.3
EUR
APOSOR907072-1G
(2,6-Dibromopyridin-4-yl)boronic acid > 98%
(2,6-Dibromopyridin-4-yl)boronic acid
Formel:
C₅H₄BBr₂NO₂ Molecular Weight: 280,71 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD18910899 CAS-Nummer: 2096340-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...