N-Methoxy-N-methylbenzamide 98%

Lieferant: Apollo Scientific

OR907094-100G
APOSOR907094-100GEA 180 EUR
APOSOR907094-100G
N-Methoxy-N-methylbenzamide 98%
N-Methoxy-N-methylbenzamide
Formel: C₉H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00075320
CAS-Nummer: 6919-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR