N-Cyclopentylpiperidine-4-carboxamide 95%
Lieferant: Apollo Scientific
OR907141-1G
APOSOR907141-1GEA
574
EUR
APOSOR907141-1G
N-Cyclopentylpiperidine-4-carboxamide 95%
N-Cyclopentylpiperidine-4-carboxamide
Formel:
C₁₁H₂₀N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817263 CAS-Nummer: 946744-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...