2,2'-Dibrombiphenyl (PBB Nr. 4) 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Brom-2-(2-bromphenyl)benzol

OR907193-10G OR907193-25G
APOSOR907193-10GEA 116 EUR
APOSOR907193-10G APOSOR907193-25G
2,2'-Dibrombiphenyl (PBB Nr. 4) 98%
2,2'-Dibrombiphenyl (PBB Nr. 4)
Formel: C₁₂H₈Br₂
Molecular Weight: 312 g/mol
Siedepunkt: 252…274 °C
Schmelzpunkt: 78…83 °C
Dichte: 1.81
MDL: MFCD00093707
CAS-Nummer: 13029-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR