2,2'-Dibrombiphenyl (PBB Nr. 4) 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-2-(2-bromphenyl)benzol
OR907193-10G
OR907193-25G
APOSOR907193-10GEA
116
EUR
APOSOR907193-10G
APOSOR907193-25G
2,2'-Dibrombiphenyl (PBB Nr. 4) 98%
2,2'-Dibrombiphenyl (PBB Nr. 4)
Formel:
C₁₂H₈Br₂ Molecular Weight: 312 g/mol Siedepunkt: 252…274 °C Schmelzpunkt: 78…83 °C Dichte: 1.81 |
MDL:
MFCD00093707 CAS-Nummer: 13029-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...