4-Morpholin-4-yl-1,2,5-thiadiazol-3-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Morpholino-1,2,5-thiadiazol-3-ol

OR9072-1G
APOSOR9072-1GEA 212 EUR
APOSOR9072-1G
4-Morpholin-4-yl-1,2,5-thiadiazol-3-ol
4-Morpholin-4-yl-1,2,5-thiadiazol-3-ol
Formel: C₆H₉N₃O₂S
Molecular Weight: 187,22 g/mol
Schmelzpunkt: 196…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01820806
CAS-Nummer: 30165-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR