Methyl-L-Isoleucinat-Hydrochlorid 97%

Lieferant: Apollo Scientific

OR907205-100G OR907205-25G OR907205-500G
APOSOR907205-100GEA 106 EUR
APOSOR907205-100G APOSOR907205-25G APOSOR907205-500G
Methyl-L-Isoleucinat-Hydrochlorid 97%
L-Isoleucine methyl ester hydrochloride
Formel: C₇H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00038911
CAS-Nummer: 18598-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR