N-Butyl-4-piperidinecarboxamide hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: N-Butylpiperidine-4-carboxamide

OR907232-1G
APOSOR907232-1GEA 502 EUR
APOSOR907232-1G
N-Butyl-4-piperidinecarboxamide hydrochloride 95%
N-Butyl-4-piperidinecarboxamide hydrochloride
Formel: C₁₀H₂₁ClN₂O
Molecular Weight: 220,74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11615740
CAS-Nummer: 73415-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR