N-Benzyl-3-chloro-propionamide 95%

Lieferant: Apollo Scientific

OR907290-1G
APOSOR907290-1GEA 342 EUR
APOSOR907290-1G
N-Benzyl-3-chloro-propionamide 95%
N-Benzyl-3-chloro-propionamide
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197.661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00018975
CAS-Nummer: 501-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR