6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR907336-5G
APOSOR907336-5GEA 172 EUR
APOSOR907336-5G
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formel: C₉H₁₁BrClN
Molecular Weight: 248.5473 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234662
CAS-Nummer: 215798-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR