N-Isopropyl-4-piperidinecarboxamide hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
N-Isopropylpiperidine-4-carboxamide
OR907356-1G
APOSOR907356-1GEA
400
EUR
APOSOR907356-1G
N-Isopropyl-4-piperidinecarboxamide hydrochloride 95%
N-Isopropyl-4-piperidinecarboxamide hydrochloride
Formel:
C₉H₁₉ClN₂O Molecular Weight: 206,72 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11615741 CAS-Nummer: 102125-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...