N-tert-Butylpiperidine-4-carboxamide 98%

Lieferant: Apollo Scientific

OR907366-1G OR907366-250MG
APOSOR907366-1GEA 574 EUR
APOSOR907366-1G APOSOR907366-250MG
N-tert-Butylpiperidine-4-carboxamide 98%
N-tert-Butylpiperidine-4-carboxamide
Formel: C₁₀H₂₀N₂O
Molecular Weight: 184.279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739717
CAS-Nummer: 86542-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR