1-Amino-4-methylcyclohexane-1-carbonitrile hydrochloride 98%
Lieferant: Apollo Scientific
OR907401-1G
APOSOR907401-1GEA
364
EUR
APOSOR907401-1G
1-Amino-4-methylcyclohexane-1-carbonitrile hydrochloride 98%
1-Amino-4-methylcyclohexane-1-carbonitrile hydrochloride
Formel:
C₈H₁₄N₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD22689078 CAS-Nummer: 92334-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...