2-Cyclohexylvinylboronic acid 97%

Lieferant: Apollo Scientific

OR907446-1G
APOSOR907446-1GEA 192 EUR
APOSOR907446-1G
2-Cyclohexylvinylboronic acid 97%
2-Cyclohexylvinylboronic acid
Formel: C₈H₁₅BO₂
Schmelzpunkt: 106…111 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01074615
CAS-Nummer: 37490-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR