2-Isopropoxy-5-methylphenylboronic acid 95%

Lieferant: Apollo Scientific

OR907454-1G
APOSOR907454-1GEA 132 EUR
APOSOR907454-1G
2-Isopropoxy-5-methylphenylboronic acid 95%
2-Isopropoxy-5-methylphenylboronic acid
Formel: C₁₀H₁₅BO₃
Molecular Weight: 194,035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04974067
CAS-Nummer: 480438-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR