5-Methyl-2-propoxyphenylboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
(5-Methyl-2-propoxyphenyl)boronic acid
OR907457-1G
APOSOR907457-1GEA
112
EUR
APOSOR907457-1G
5-Methyl-2-propoxyphenylboronic acid 95%
5-Methyl-2-propoxyphenylboronic acid
Formel:
C₁₀H₁₅BO₃ Molecular Weight: 194.035 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974066 CAS-Nummer: 480438-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...