5-Isopropyl-2-methoxybenzeneboronic acid 95%

Lieferant: Apollo Scientific

Synonyme: (5-Isopropyl-2-methoxyphenyl)boronic acid

OR907465-1G
APOSOR907465-1GEA 77 EUR
APOSOR907465-1G
5-Isopropyl-2-methoxybenzeneboronic acid 95%
5-Isopropyl-2-methoxybenzeneboronic acid
Formel: C₁₀H₁₅BO₃
Molecular Weight: 194,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318154
CAS-Nummer: 216393-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR