1-Propylpyrazole-4-boronic acid 97%

Lieferant: Apollo Scientific

Synonyme: (1-Propyl-1H-pyrazol-4-yl)boronic acid

OR907482-5G
APOSOR907482-5GEA 186 EUR
APOSOR907482-5G
1-Propylpyrazole-4-boronic acid 97%
1-Propylpyrazole-4-boronic acid
Formel: C₆H₁₁BN₂O₂
Molecular Weight: 153.975 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11504862
CAS-Nummer: 847818-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR