5-Acetyl-2-methoxyphenylboronic acid 95%

Lieferant: Apollo Scientific

OR907499-1G
APOSOR907499-1GEA 36.4 EUR
APOSOR907499-1G
5-Acetyl-2-methoxyphenylboronic acid 95%
5-Acetyl-2-methoxyphenylboronic acid
Formel: C₉H₁₁BO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09839106
CAS-Nummer: 1215281-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR