(1-Benzothiophen-4-yl)boronic acid 98%

Lieferant: Apollo Scientific

OR907505-1G
APOSOR907505-1GEA 0 EUR
APOSOR907505-1G
(1-Benzothiophen-4-yl)boronic acid 98%
(1-Benzothiophen-4-yl)boronic acid
Formel: C₈H₇BO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD22191951
CAS-Nummer: 177735-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR