1,4-Dioxa-8-azaspiro[4.5]decane hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR907516-100G
OR907516-25G
APOSOR907516-100GEA
78.3
EUR
APOSOR907516-100G
APOSOR907516-25G
1,4-Dioxa-8-azaspiro[4.5]decane hydrochloride 97%
1,4-Dioxa-8-azaspiro[4.5]decane hydrochloride
Formel:
C₇H₁₄ClNO₂ Molecular Weight: 179.6446 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791123 CAS-Nummer: 42899-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...