1,4-Dioxa-8-azaspiro[4.5]decane hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR907516-100G OR907516-25G
APOSOR907516-100GEA 78.3 EUR
APOSOR907516-100G APOSOR907516-25G
1,4-Dioxa-8-azaspiro[4.5]decane hydrochloride 97%
1,4-Dioxa-8-azaspiro[4.5]decane hydrochloride
Formel: C₇H₁₄ClNO₂
Molecular Weight: 179.6446 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03791123
CAS-Nummer: 42899-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR