2-Ethoxyisonicotinsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-Ethoxypyridine-4-carboxylic acid

OR907519-25G
APOSOR907519-25GEA 626 EUR
APOSOR907519-25G
2-Ethoxyisonicotinsäure 98%
2-Ethoxyisonicotinsäure
Formel: C₈H₉NO₃
Molecular Weight: 167.1620 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115719
CAS-Nummer: 91940-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR